missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Phenylglutaric acid, 98%

Manufacturer:  Alfa Aesar™ A1457106

 View more versions of this product

Catalog No. AAA1457106



Specifications

Specifications

3-Phenylglutaric acid
5g
C11H12O4
2052171
RZOKZOYSUCSPDF-UHFFFAOYSA-N
3-phenylpentanedioic acid
77823
98%
140°C to 143°C
4165-96-2
MFCD00002718
3-phenylglutaric acid, pentanedioic acid, 3-phenyl, 3-phenyl-pentanedioic acid, acmc-20amyr, #, 3-phenylglutaric acid
C1=CC=C(C=C1)C(CC(=O)O)CC(=O)O
208.213
208.21

Chemical Identifiers

4165-96-2
208.213
RZOKZOYSUCSPDF-UHFFFAOYSA-N
77823
C1=CC=C(C=C1)C(CC(=O)O)CC(=O)O
C11H12O4
MFCD00002718
3-phenylglutaric acid, pentanedioic acid, 3-phenyl, 3-phenyl-pentanedioic acid, acmc-20amyr, #, 3-phenylglutaric acid
3-phenylpentanedioic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents