Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(Phenylaminomethyl)benzeneboronic acid pinacol ester, 97%

$76.19 - $76.19

Chemical Identifiers

Chemical Name or Material 3-(Phenylaminomethyl)benzeneboronic acid pinacol ester
Molecular Formula C19H24BNO2
Formula Weight 309.21
MDL Number MFCD09266198
Synonym 3-(Phenylaminomethyl)phenylboronic acid pinacol ester; N-[3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)]benzyl]aniline
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2828706

 
alfa aesar™
H2828706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH2828703

 
alfa aesar™
H2828703
1g Each for $76.19
Description & Specifications

Specifications

Formula Weight 309.21
Chemical Name or Material 3-(Phenylaminomethyl)benzeneboronic acid pinacol ester
Assay 97%
Molecular Formula C19H24BNO2
MDL Number MFCD09266198
Synonym 3-(Phenylaminomethyl)phenylboronic acid pinacol ester; N-[3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)]benzyl]aniline
Melting Point 112° to 115°C
Quantity 5g