missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-Phenylacrylaldehyde oxime, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals BTB09548EB
Specifications
3-Phenylacrylaldehyde oxime | |
97% | |
MFCD00019969 | |
cinnamaldehyde oxime, 3-phenylacrylaldehyde oxime, phenylacrylaldehyde oxime, e-n-2e-3-phenylprop-2-en-1-ylidene hydroxylamine, n-hydroxy-3-phenylprop-2-en-1-imine, n-3-phenylprop-2-en-1-ylidene hydroxylamine, 3-phenylpropenal oxime, benzeneacrylaldehydeoxime, 3-phenyl-2-propenal oxime, cinnamaldoxime, e + z | |
O\N=C\C=C\C1=CC=CC=C1 | |
147.18 | |
147.18 |
13372-81-1 | |
C9H9NO | |
25g | |
RUQDOYIAKHIMAN-DAAQNPAKSA-N | |
(NE)-N-[(E)-3-phenylprop-2-enylidene]hydroxylamine | |
9561350 |
Chemical Identifiers
13372-81-1 | |
147.18 | |
RUQDOYIAKHIMAN-DAAQNPAKSA-N | |
9561350 | |
O\N=C\C=C\C1=CC=CC=C1 |
C9H9NO | |
MFCD00019969 | |
cinnamaldehyde oxime, 3-phenylacrylaldehyde oxime, phenylacrylaldehyde oxime, e-n-2e-3-phenylprop-2-en-1-ylidene hydroxylamine, n-hydroxy-3-phenylprop-2-en-1-imine, n-3-phenylprop-2-en-1-ylidene hydroxylamine, 3-phenylpropenal oxime, benzeneacrylaldehydeoxime, 3-phenyl-2-propenal oxime, cinnamaldoxime, e + z | |
(NE)-N-[(E)-3-phenylprop-2-enylidene]hydroxylamine |