Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Phenyl-5-isoxazolone, 98%

$52.36 - $358.15

Chemical Identifiers

Chemical Name or Material 3-Phenyl-5-isoxazolone
CAS 1076-59-1
Molecular Formula C9H7NO2
Formula Weight 161.16g/mol
MDL Number MFCD00003158
Beilstein 5075
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1583706

 
alfa aesar™
A15837-06
5g Each for $52.36

AAA1583714

 
alfa aesar™
A15837-14
25g Each for $129.35

AAA1583722

 
alfa aesar™
A15837-22
100g Each for $358.15
Description & Specifications

Specifications

Melting Point 150° to 154°C
Quantity 5g
Chemical Name or Material 3-Phenyl-5-isoxazolone
CAS 1076-59-1
Assay 98%
Molecular Formula C9H7NO2
Formula Weight 161.16g/mol
MDL Number MFCD00003158
Beilstein 5075
TSCA TSCA