Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Phenyl-4(3H)-quinazolinone, 98%

$40.47 - $632.65

Chemical Identifiers

Chemical Name or Material 3-Phenyl-4(3H)-quinazolinone
CAS 16347-60-7
Molecular Formula C14H10N2O
Formula Weight 222.25
MDL Number MFCD00024102
Synonym 4-Keto-3-phenyl-3,4-dihydroquinazoline
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1815206

  SDS
alfa aesar™
A1815206
5g Each for $157.00

AAA1815203

  SDS
alfa aesar™
A1815203
1g Each for $40.47

AAA1815214

  SDS
alfa aesar™
A1815214
25g Each for $632.65
Description & Specifications

Specifications

Formula Weight 222.25
Chemical Name or Material 3-Phenyl-4(3H)-quinazolinone
Assay 98%
Molecular Formula C14H10N2O
MDL Number MFCD00024102
CAS 16347-60-7
Synonym 4-Keto-3-phenyl-3,4-dihydroquinazoline
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 138° to 140°C
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.