Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Phenyl-3-pentanol, 97%

$200.41 - $798.58

Chemical Identifiers

Chemical Name or Material 3-Phenyl-3-pentanol
CAS 1565-71-5
Molecular Formula C11H16O
Formula Weight 164.25
MDL Number MFCD00021830
Synonym Diethyl phenyl carbinol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2087114

  SDS
alfa aesar™
B2087114
25g Each for $798.58

AAB2087106

  SDS
alfa aesar
B2087106
5g Each for $200.41
Description & Specifications

Specifications

Formula Weight 164.25
Chemical Name or Material 3-Phenyl-3-pentanol
Assay 97%
Molecular Formula C11H16O
MDL Number MFCD00021830
CAS 1565-71-5
Synonym Diethyl phenyl carbinol
Boiling Point 98° to 100°C (3mmHg)
Quantity 25g

Applications
It finds its application in the synthesis and physical properties of some c12 and c13 phenylalkane and cyclohexylalkane hydrocarbons and in the dehydration of Alcohols in Dimethyl Sulfoxide1,2.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.