Learn More
3-Phenyl-2-propyn-1-ol, 98%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC270030050
Specifications
3-Phenyl-2-propyn-1-ol | |
>110°C | |
Glass bottle | |
1.078 | |
Colorless to Yellow | |
1504-58-1 | |
100.0 | |
C9H8O | |
MFCD00040914 | |
NITUNGCLDSFVDL-UHFFFAOYSA-N | |
3-phenylprop-2-yn-1-ol | |
123115 | |
Liquid |
1.0780g/mL | |
Authentic | |
1.5820 to 1.5840 | |
129.0°C to 130.0°C (10.0mmHg) | |
5g | |
97.5 | |
97.5% min. (GC) | |
C6H5C≡CCH2OH | |
1-hydroxymethyl-2-phenylacetylene, 1-phenyl-1-propyn-3-ol, 1-phenyl-propyn-3-ol, 2-propyn-1-ol, 3-phenyl, 3-phenyl-2-propyn-1-ol, 3-phenyl-2-propyn1-ol, 3-phenyl-prop-2-yn-1-ol, 3-phenylpropargyl alcohol, acmc-1bs1u, pubchem13580 | |
C1=CC=C(C=C1)C#CCO | |
132.16 | |
132.16 | |
98% |
Chemical Identifiers
1504-58-1 | |
132.16 | |
NITUNGCLDSFVDL-UHFFFAOYSA-N | |
123115 | |
C1=CC=C(C=C1)C#CCO |
C9H8O | |
MFCD00040914 | |
1-hydroxymethyl-2-phenylacetylene, 1-phenyl-1-propyn-3-ol, 1-phenyl-propyn-3-ol, 2-propyn-1-ol, 3-phenyl, 3-phenyl-2-propyn-1-ol, 3-phenyl-2-propyn1-ol, 3-phenyl-prop-2-yn-1-ol, 3-phenylpropargyl alcohol, acmc-1bs1u, pubchem13580 | |
3-phenylprop-2-yn-1-ol |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning