Alfa Aesar™ 3-Phenyl-2-propyn-1-ol, 98+%
Manufacturer:  Alfa Aesar™ 4377809
 View more versions of this product
Catalog No. AA4377809
Specifications
Specifications
3-Phenyl-2-propyn-1-ol
>110°C (230°F)
129°C to 130°C (10mmHg)
1504-58-1
MFCD00040914
3-phenyl-2-propyn-1-ol, 2-propyn-1-ol, 3-phenyl, 3-phenyl-prop-2-yn-1-ol, 1-hydroxymethyl-2-phenylacetylene, 1-phenyl-1-propyn-3-ol, pubchem13580, 3-phenyl-2-propyn1-ol, 3-phenylpropargyl alcohol, acmc-1bs1u, 1-phenyl-propyn-3-ol
C1=CC=C(C=C1)C#CCO
132.162
132.16
≥98%
1.078g/mL
1.5867
10g
C9H8O
2040911
NITUNGCLDSFVDL-UHFFFAOYSA-N
3-phenylprop-2-yn-1-ol
123115
Liquid
Chemical Identifiers
1504-58-1
132.162
NITUNGCLDSFVDL-UHFFFAOYSA-N
123115
C1=CC=C(C=C1)C#CCO
C9H8O
MFCD00040914
3-phenyl-2-propyn-1-ol, 2-propyn-1-ol, 3-phenyl, 3-phenyl-prop-2-yn-1-ol, 1-hydroxymethyl-2-phenylacetylene, 1-phenyl-1-propyn-3-ol, pubchem13580, 3-phenyl-2-propyn1-ol, 3-phenylpropargyl alcohol, acmc-1bs1u, 1-phenyl-propyn-3-ol
3-phenylprop-2-yn-1-ol
Documents