Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Phenyl-2-piperazinone, 97%

Chemical Identifiers

Chemical Name or Material 3-Phenyl-2-piperazinone
CAS 5368-28-5
Formula Weight 176.22
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6309903

 
alfa aesar™
H6309903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6309906

 
alfa aesar™
H6309906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH63099MD

 
alfa aesar™
H63099MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 176.22
Chemical Name or Material 3-Phenyl-2-piperazinone
Physical Form Solid
Quantity 1g
Assay 97%
CAS 5368-28-5