Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Phenyl-2-oxazolidinone, 98%

$94.93 - $94.93

Chemical Identifiers

Chemical Name or Material 3-Phenyl-2-oxazolidinone
CAS 703-56-0
Molecular Formula C9H9NO2
Formula Weight 163.18
MDL Number MFCD00020870
Beilstein 139712
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0848406

  SDS
alfa aesar™
L0848406
5g Each for $94.93
Description & Specifications

Specifications

Formula Weight 163.18
CAS 703-56-0
Assay 98%
Chemical Name or Material 3-Phenyl-2-oxazolidinone
Molecular Formula C9H9NO2
MDL Number MFCD00020870
Melting Point 120° to 122°C
Beilstein 139712
Quantity 5g