Please login to your online account to display your discounted pricing

3-Phenyl-1H-pyrazol-5-amine, 97%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  KM00060DA

 View more versions of this product

Catalog No. KM00060DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 159.19
Quantity 1g
Assay 97%
Chemical Name or Material 5-Phenyl-1H-pyrazol-3-amine
Assay Percent Range 97%
Molecular Formula C9H9N3
Packaging Amber Glass Bottle
CAS 1572-10-7
Health Hazard 1 Warning

Specifications

Chemical Name or Material 5-Phenyl-1H-pyrazol-3-amine
Assay 97%
Assay Percent Range 97%
Molecular Formula C9H9N3
Formula Weight 159.19
Accessories

Warning

SDS & Safety Documents