Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Phenyl-1-propyl acetate, 98%

$129.27 - $449.01

Chemical Identifiers

Chemical Name or Material 3-Phenyl-1-propyl acetate
CAS 122-72-5
Molecular Formula CH3CO2(CH2)3C6H5
Formula Weight 178.23
MDL Number MFCD00026216
Synonym 1-Acetoxy-3-phenylpropane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2087818

  SDS
alfa aesar™
B2087818
50g Each for $129.27

AAB2087830

  SDS
alfa aesar™
B2087830
250g Each for $449.01
Description & Specifications

Specifications

Formula Weight 178.23
Chemical Name or Material 3-Phenyl-1-propyl acetate
TSCA TSCA
Assay 98%
Molecular Formula CH3CO2(CH2)3C6H5
MDL Number MFCD00026216
CAS 122-72-5
Synonym 1-Acetoxy-3-phenylpropane
RTECS Number UB9000000
Flash Point 130°C (266°F)
Boiling Point 252°C
Quantity 50g