Please login to your online account to display your discounted pricing

3-Phenoxytoluene, 98+%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 3-Phenoxytoluene
CAS 3586-14-9
CAS Min % 98.0
CAS Max % 100.0
Assay ≥98.0%
Assay Percent Range 98+%
Molecular Formula C13H12O
Linear Formula C6H5OC6H4CH3
Formula Weight 184.24
MDL Number MFCD00008531
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC185101000

  SDS
fisher scientific acros
185101000
Glass bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Assay ≥98.0%
Formula Weight 184.24
Quantity 100g
Color Clear very slightly yellow
Density 1.0500g/mL
Beilstein 06, 377
CAS Min % 98.0
CAS Max % 100.0
Chemical Name or Material 3-Phenoxytoluene
Refractive Index 1.5720 to 1.5740 (20°C, 589nm)
Assay Percent Range 98+%
Molecular Formula C13H12O
CAS 3586-14-9
Synonym Phenyl m-tolyl ether
Packaging Glass bottle
Linear Formula C6H5OC6H4CH3
MDL Number MFCD00008531
Infrared Spectrum Authentic