Please login to your online account to display your discounted pricing

3-Phenoxytoluene, 98+%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: fisher scientific acros  185101000

 View more versions of this product

Catalog No. AC185101000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.0%
Formula Weight 184.24
Physical Form Liquid
Quantity 100g
Color Clear very slightly yellow
Density 1.0500g/mL
Beilstein 06, 377
CAS Min % 98.0
CAS Max % 100.0
Synonym Phenyl m-tolyl ether
Assay Percent Range 98+%
Refractive Index 1.5720 to 1.5740 (20°C, 589nm)
CAS 3586-14-9
Molecular Formula C13H12O
Chemical Name or Material 3-Phenoxytoluene
Packaging Glass bottle
Infrared Spectrum Authentic
Linear Formula C6H5OC6H4CH3
MDL Number MFCD00008531

Specifications

Chemical Name or Material 3-Phenoxytoluene
CAS 3586-14-9
CAS Min % 98.0
CAS Max % 100.0
Assay ≥98.0%
Assay Percent Range 98+%
Molecular Formula C13H12O
Linear Formula C6H5OC6H4CH3
Formula Weight 184.24
MDL Number MFCD00008531
Synonym Phenyl m-tolyl ether