missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Phenoxythiophene, 97%

$119.36 - $2,091.30

Chemical Identifiers

CAS 63285-84-7
Molecular Formula C10H8OS
Formula Weight 176.23
MDL Number MFCD00178747
Chemical Name or Material 3-Phenoxythiophene
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2528803 SDS Alfa Aesar™
B2528803
1g Each for $119.36
AAB2528806 SDS Alfa Aesar™
B2528806
5g Each for $523.57
AAB2528814 SDS Alfa Aesar™
B2528814
25g Each for $2,091.30
Specifications Description & Specifications

Chemical Identifiers

63285-84-7
176.23
3-Phenoxythiophene
C10H8OS
MFCD00178747

Specifications

1g
C10H8OS
MFCD00178747
97%
63285-84-7
176.23
3-Phenoxythiophene

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger