Please login to your online account to display your discounted pricing

3-Phenoxypropyl Bromide 95%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: fisher scientific acros  130260250

 View more versions of this product

Catalog No. AC130260250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Assay ≥94%
Formula Weight 215.09
Quantity 25g
Color Clear slightly brown
EINECS Number 209-623-4
TSCA TSCA
Density 1.3600g/mL
Beilstein 06, 142
CAS Min % 94.0
CAS Max % 100.0
Chemical Name or Material 3-Phenoxypropyl bromide
Packaging Glass bottle
Refractive Index 1.5450 to 1.5470 (20°C, 589nm)
Name Note 95%
Assay Percent Range 95%
Molecular Formula C9H11BrO
Linear Formula C6H5O(CH2)3Br
MDL Number MFCD00000256
CAS 588-63-6
Synonym 3-Bromopropyl phenyl ether
Infrared Spectrum Authentic

Specifications

Chemical Name or Material 3-Phenoxypropyl bromide
Name Note 95%
CAS 588-63-6
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C9H11BrO
Linear Formula C6H5O(CH2)3Br
Formula Weight 215.09
MDL Number MFCD00000256
Synonym 3-Bromopropyl phenyl ether