Please login to your online account to display your discounted pricing

3-Phenoxypropyl Bromide 95%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 3-Phenoxypropyl bromide
Name Note 95%
CAS 588-63-6
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C9H11BrO
Linear Formula C6H5O(CH2)3Br
Formula Weight 215.09
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC130260250

  SDS
fisher scientific acros
130260250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Assay ≥94%
Formula Weight 215.09
Quantity 25g
Color Clear slightly brown
EINECS Number 209-623-4
TSCA TSCA
Density 1.3600g/mL
Beilstein 06, 142
CAS Min % 94.0
CAS Max % 100.0
Chemical Name or Material 3-Phenoxypropyl bromide
Packaging Glass bottle
Refractive Index 1.5450 to 1.5470 (20°C, 589nm)
Name Note 95%
Assay Percent Range 95%
Molecular Formula C9H11BrO
Linear Formula C6H5O(CH2)3Br
MDL Number MFCD00000256
CAS 588-63-6
Synonym 3-Bromopropyl phenyl ether
Infrared Spectrum Authentic