Please login to your online account to display your discounted pricing

3-Phenoxypropionic acid, 99%, ACROS Organics™

$272.29 - $272.29

Chemical Identifiers

Chemical Name or Material 3-Phenoxypropionic acid
Name Note 99%
CAS 7170-38-9
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C9H10O3
Linear Formula C6H5OCH2CH2CO2H
Formula Weight 166.18
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC130250250

  SDS
thermo fisher scientific
130250250
Glass bottle 25g Each for $272.29
Description & Specifications

Specifications

Assay ≥98.5%
Formula Weight 166.18
Physical Form Fluffy Powder
Quantity 25g
Color White to beige
TSCA TSCA
EINECS Number 230-518-4
Melting Point 96° to 100°C
Beilstein 06, 163
CAS Max % 100.0
CAS Min % 98.5
Infrared Spectrum Authentic
Packaging Glass bottle
Assay Percent Range 99%
Molecular Formula C9H10O3
Linear Formula C6H5OCH2CH2CO2H
Name Note 99%
Chemical Name or Material 3-Phenoxypropionic acid
CAS 7170-38-9
MDL Number MFCD00002769