Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Phenoxyphenylacetonitrile, 98%

$60.03 - $60.03

Chemical Identifiers

Chemical Name or Material 3-Phenoxyphenylacetonitrile
CAS 51632-29-2
Molecular Formula C14H11NO
Formula Weight 209.25
MDL Number MFCD00016393
Beilstein 2106883
Synonym 3-Phenoxybenzyl cyanide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0991603

 
alfa aesar™
L0991603
1g Each for $60.03
Description & Specifications

Specifications

Formula Weight 209.25
Molecular Formula C14H11NO
MDL Number MFCD00016393
Beilstein 2106883
Synonym 3-Phenoxybenzyl cyanide
Health Hazard 3 GHS P Statement
P280f
Wear protective clothing.
Chemical Name or Material 3-Phenoxyphenylacetonitrile
Assay 98%
CAS 51632-29-2
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302
Harmful if swallowed.
RTECS Number AM1370000
Flash Point >110°C (230°F)
Boiling Point 137°C (0.3mmHg)
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P280f
Wear protective clothing.

GHS H Statement
H302
Harmful if swallowed.