missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-(Phenoxymethyl)benzaldehyde, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC63604DA

 View more versions of this product

Catalog No. CC63604DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

3-(Phenoxymethyl)benzaldehyde
96.5
97%
C14H12O2
1g
KZUJGQAFTCDBKA-UHFFFAOYSA-N
3-(phenoxymethyl)benzaldehyde
21288517
97%
104508-27-2
100.0
Amber Glass Bottle
MFCD09025913
3-phenoxymethyl benzaldehyde, benzaldehyde,3-phenoxymethyl, acmc-1cada
C1=CC=C(C=C1)OCC2=CC=CC(=C2)C=O
212.248
212.25

Chemical Identifiers

104508-27-2
212.248
KZUJGQAFTCDBKA-UHFFFAOYSA-N
21288517
C1=CC=C(C=C1)OCC2=CC=CC(=C2)C=O
C14H12O2
MFCD09025913
3-phenoxymethyl benzaldehyde, benzaldehyde,3-phenoxymethyl, acmc-1cada
3-(phenoxymethyl)benzaldehyde
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions