missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Phenoxybenzyl chloride, 97%

Manufacturer:  Alfa Aesar™ A1567014

 View more versions of this product

Catalog No. AAA1567014



Specifications

Specifications

3-Phenoxybenzyl chloride
141°C to 142°C (5mmHg)
53874-66-1
MFCD00040866
2212101
QUYVTGFWFHQVRO-UHFFFAOYSA-N
1-(chloromethyl)-3-phenoxybenzene
93291
97%
>110°C (230°F)
25g
C13H11ClO
UN3265
3-phenoxybenzyl chloride, 1-chloromethyl-3-phenoxybenzene, m-phenoxybenzyl chloride, benzene, 1-chloromethyl-3-phenoxy, 3-phenoxyphenylmethyl chloride, 3-phenoxybenzylchloride, 3-chloromethyl-1-phenoxybenzene, 1-chlormethyl-3-phenoxybenzol, acmc-1asdb, m-phenoxy-benzyl chloride
C1=CC=C(C=C1)OC2=CC=CC(=C2)CCl
218.68
218.69

Chemical Identifiers

53874-66-1
218.68
QUYVTGFWFHQVRO-UHFFFAOYSA-N
93291
C1=CC=C(C=C1)OC2=CC=CC(=C2)CCl
C13H11ClO
MFCD00040866
3-phenoxybenzyl chloride, 1-chloromethyl-3-phenoxybenzene, m-phenoxybenzyl chloride, benzene, 1-chloromethyl-3-phenoxy, 3-phenoxyphenylmethyl chloride, 3-phenoxybenzylchloride, 3-chloromethyl-1-phenoxybenzene, 1-chlormethyl-3-phenoxybenzol, acmc-1asdb, m-phenoxy-benzyl chloride
1-(chloromethyl)-3-phenoxybenzene
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

RTECSNumber : CZ0790000

Documents