Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Phenoxybenzyl alcohol, 98%

$148.72 - $623.80

Chemical Identifiers

Chemical Name or Material 3-Phenoxybenzyl alcohol
CAS 13826-35-2
Molecular Formula C13H12O2
Formula Weight 200.24
MDL Number MFCD00004636
Beilstein 475312
Synonym 3-(Hydroxymethyl)diphenyl ether
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1300818

  SDS
alfa aesar™
A1300818
50g Each for $148.72

AAA1300830

  SDS
alfa aesar™
A1300830
250g Each for $623.80
Description & Specifications

Specifications

Formula Weight 200.24
Chemical Name or Material 3-Phenoxybenzyl alcohol
TSCA TSCA
Assay 98%
Molecular Formula C13H12O2
MDL Number MFCD00004636
CAS 13826-35-2
Beilstein 475312
Synonym 3-(Hydroxymethyl)diphenyl ether
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Boiling Point 300°C
Melting Point 4° to 7°C
Flash Point 186°C (366°F)
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number DP0775400
Quantity 50g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.