Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Phenoxybenzoic acid, 99%

$44.45 - $378.20

Chemical Identifiers

Chemical Name or Material 3-Phenoxybenzoic acid
CAS 3739-38-6
Molecular Formula C13H10O3
Formula Weight 214.22
MDL Number MFCD00002498
Beilstein 2105574
Synonym 3-Carboxydiphenyl ether
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1483106

  SDS
alfa aesar™
A1483106
5g Each for $44.45

AAA1483114

  SDS
alfa aesar™
A1483114
25g Each for $145.54

AAA1483122

  SDS
alfa aesar™
A1483122
100g Each for $378.20
Description & Specifications

Specifications

Formula Weight 214.22
Assay 99%
Molecular Formula C13H10O3
Synonym 3-Carboxydiphenyl ether
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
RTECS Number DH6293500
Quantity 5g
Chemical Name or Material 3-Phenoxybenzoic acid
MDL Number MFCD00002498
CAS 3739-38-6
Beilstein 2105574
Health Hazard 1 Warning
Melting Point 146° to 149°C
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Applications
3-Phenoxybenzoic acid are used to control a wide range of insects in public and commercial buildings, animal facilities, warehouses, agricultural fields, and greenhouses. They are also applied on livestock to control insects. In agriculture, cypermethrin, cyfluthrin, and deltamethrin have been used frequently on cotton. It is generally formulated as complex mixtures of different chemical isomers, solvent oils, and synergists.

Solubility
It is soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under normal temperatures and pressures. Keep away from strong oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.