Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Phenoxybenzaldehyde, 98%

Chemical Identifiers

Chemical Name or Material 3-Phenoxybenzaldehyde
CAS 39515-51-0
Molecular Formula C13H10O2
Formula Weight 198.22
MDL Number MFCD00003353
UN Number UN2810
Beilstein 511662
Synonym 3-Formyldiphenyl ether
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1378018

  SDS
alfa aesar™
A1378018
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1378009

  SDS
alfa aesar™
A1378009
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1378030

  SDS
alfa aesar™
A1378030
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 198.22
Chemical Name or Material 3-Phenoxybenzaldehyde
TSCA TSCA
Assay 98%
Molecular Formula C13H10O2
MDL Number MFCD00003353
CAS 39515-51-0
UN Number UN2810
Beilstein 511662
Synonym 3-Formyldiphenyl ether
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H330
Toxic if swallowed.
Fatal if inhaled.
Boiling Point 139° to 140°C (0.1mmHg)
Melting Point 13°C
Flash Point 186°C (366°F)
Health Hazard 3 GHS P Statement
P260-P301+P310-P304+P340-P320-P330-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
RTECS Number CU7560200
Quantity 50g
Safety and Handling

Danger

GHS P Statement
P260-P301+P310-P304+P340-P320-P330-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H330
Toxic if swallowed.
Fatal if inhaled.