Please login to your online account to display your discounted pricing

3-Phenoxybenzaldehyde, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 3-Phenoxybenzaldehyde
Name Note 97%
CAS 39515-51-0
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C13H10O2
Linear Formula C6H5OC6H4CHO
Formula Weight 198.22
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC184790050

  SDS
thermo fisher scientific
184790050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 198.22
Physical Form Liquid
Assay ≥96.0%
Quantity 5g
Color Clear yellow to amber
RTECS Number CU7560200
TSCA TSCA
EINECS Number 254-487-1
Density 1.1470g/mL
CAS Min % 96.0
CAS Max % 100.0
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Refractive Index 1.5980 to 1.6000 (20°C, 589nm)
Chemical Name or Material 3-Phenoxybenzaldehyde
Infrared Spectrum Authentic
Assay Percent Range 97%
MDL Number MFCD00003353
Name Note 97%
Molecular Formula C13H10O2
CAS 39515-51-0
Linear Formula C6H5OC6H4CHO
Safety and Handling

Danger

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.