missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Phenoxy-1-propanol, TCI America™

Manufacturer:  TCI America P225425G

 View more versions of this product

Catalog No. P225425G



Specifications

Specifications

3-Phenoxy-1-propanol
Colorless
6180-61-6
MFCD00039549
AWVDYRFLCAZENH-UHFFFAOYSA-N
3-phenoxypropan-1-ol
80319
Clear Liquid at 20°C
153°C
25g
C9H12O2
3-phenoxy-1-propanol, 1-propanol, 3-phenoxy, 3-phenoxy 1-propanol, unii-515pbp1u72, propoxylated phenol, ph 4p, polypropylene glycol phenyl ether, 3-phenoxypropanol, 3-phenyloxypropanol, polypropylene glycol monophenyl ether
C1=CC=C(C=C1)OCCCO
152.193
152.19
≥98.0% (GC)

Chemical Identifiers

6180-61-6
152.193
AWVDYRFLCAZENH-UHFFFAOYSA-N
80319
C1=CC=C(C=C1)OCCCO
C9H12O2
MFCD00039549
3-phenoxy-1-propanol, 1-propanol, 3-phenoxy, 3-phenoxy 1-propanol, unii-515pbp1u72, propoxylated phenol, ph 4p, polypropylene glycol phenyl ether, 3-phenoxypropanol, 3-phenyloxypropanol, polypropylene glycol monophenyl ether
3-phenoxypropan-1-ol
Safety and Handling

Safety and Handling

TSCA : No

Documents