missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Pentyn-1-ol, 99%, ACROS Organics™

Manufacturer:  Acros Organics AC197331000

 View more versions of this product

Catalog No. AC197331000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

3-Pentyn-1-ol
54°C
1.4550 to 1.4570
154.0°C to 157.0°C
99%
98.5
99%
CH3C≡CCH2CH2OH
3-pentyn-1-ol, 3-pentynol, acmc-1bxad, 3-pentyn-1-ol
CC#CCCO
84.118
84.12
0.9100g/mL
Glass bottle
0.91
100g
10229-10-4
100.0
C5H8O
MFCD00002956
IDYNOORNKYEHHO-UHFFFAOYSA-N
pent-3-yn-1-ol
66295
99%

Chemical Identifiers

10229-10-4
84.118
IDYNOORNKYEHHO-UHFFFAOYSA-N
66295
CC#CCCO
C5H8O
MFCD00002956
3-pentyn-1-ol, 3-pentynol, acmc-1bxad, 3-pentyn-1-ol
pent-3-yn-1-ol
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Causes skin irritation.
Harmful if inhaled.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable liquid and vapor.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 233-550-7

TSCA : TSCA

Documents