Learn More
3-Pentyn-1-ol, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 197330250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
3-Pentyn-1-ol | |
10229-10-4 | |
100.0 | |
154.0°C to 157.0°C | |
98.5% min. (GC) | |
C5H8O | |
CH3C≡CCH2CH2OH | |
25 g | |
0.91 | |
IDYNOORNKYEHHO-UHFFFAOYSA-N | |
pent-3-yn-1-ol | |
66295 | |
99% |
99% | |
98.5 | |
0.9100g/mL | |
54°C | |
Glass bottle | |
1.4550 to 1.4570 | |
MFCD00002956 | |
01,IV,2227 | |
3-pentyn-1-ol, 3-pentynol, acmc-1bxad | |
CC#CCCO | |
84.12 | |
84.12 |
Chemical Identifiers
10229-10-4 | |
84.12 | |
IDYNOORNKYEHHO-UHFFFAOYSA-N | |
66295 | |
CC#CCCO |
C5H8O | |
MFCD00002956 | |
3-pentyn-1-ol, 3-pentynol, acmc-1bxad | |
pent-3-yn-1-ol |
Safety and Handling
GHS H Statement
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rin
GHS Signal Word: Warning
EINECSNumber : 233-550-7
TSCA : TSCA
RUO – Research Use Only