Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Pentyl formate, 94%

Chemical Identifiers

Chemical Name or Material 3-Pentyl formate
CAS 58368-67-5
Molecular Formula HCO2CH(CH2CH3)2
Formula Weight 116.16
MDL Number MFCD00049220
UN Number UN3272
Synonym Formic acid 3-pentyl ester; Formic acid 3-pentyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2056606

 
alfa aesar™
B2056606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2056614

 
alfa aesar™
B2056614
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 116.16
Chemical Name or Material 3-Pentyl formate
TSCA TSCA
Assay 94%
Boiling Point 116° to 118°C
Molecular Formula HCO2CH(CH2CH3)2
Quantity 5g
MDL Number MFCD00049220
CAS 58368-67-5
UN Number UN3272
Synonym Formic acid 3-pentyl ester; Formic acid 3-pentyl ester
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H226
Flammable liquid and vapour.
Health Hazard 3 GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
Safety and Handling

Warning

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226
Flammable liquid and vapour.