missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Penten-2-one, 70%, Tech., ACROS Organics™

Manufacturer:  Acros Organics AC163290050

 View more versions of this product

Catalog No. AC163290050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

3-Penten-2-one
21°C
Glass bottle
0.86
Colorless
625-33-2
100.0
70%
CH3CH=CHCOCH3
02,306; 03,218; 04,371
LABTWGUMFABVFG-ONEGZZNKSA-N
(E)-pent-3-en-2-one
637920
Liquid
0.8600g/mL
Authentic
1.4405
121.0°C to 124.0°C
5g
65.0
Technical
C5H8O
MFCD00009290
z-pent-3-en-2-one, unii-w8xx21whyc, z-3-penten-2-one, w8xx21whyc, 3-penten-2-one, 3z-pent-3-en-2-one, 3-penten-2-one, z, z-3-penten-2-one, cis-3-penten-2-one, 3-penten-2-one, cis
CC=CC(=O)C
84.118
84.12
70%

Chemical Identifiers

625-33-2
84.118
LABTWGUMFABVFG-ONEGZZNKSA-N
637920
CC=CC(=O)C
C5H8O
MFCD00009290
z-pent-3-en-2-one, unii-w8xx21whyc, z-3-penten-2-one, w8xx21whyc, 3-penten-2-one, 3z-pent-3-en-2-one, 3-penten-2-one, z, z-3-penten-2-one, cis-3-penten-2-one, 3-penten-2-one, cis
(E)-pent-3-en-2-one
Safety and Handling

Safety and Handling

GHS H Statement:
Toxic in contact with skin.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Highly flammable liquid and vapour.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS Documents