Please login to your online account to display your discounted pricing

3-Penten-2-one, 70%, Tech., ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC163290010

 View more versions of this product

Catalog No. AC163290010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Purity Grade Technical Grade
Physical Form Liquid
Assay ≥70.0%
Formula Weight 84.12
Quantity 1g
Color Clear colorless
Density 0.86g/mL
Fieser 02,306; 03,218; 04,371
CAS Min % 65.0
CAS Max % 100.0
Chemical Name or Material 3-Penten-2-one
MDL Number MFCD00009290
Health Hazard 2 GHS H Statement:
Toxic in contact with skin.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Highly flammable liquid and vapour.
Infrared Spectrum Authentic
Assay Percent Range 70%
Molecular Formula C5H8O
Linear Formula CH3CH=CHCOCH3
CAS 625-33-2
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle

Specifications

Chemical Name or Material 3-Penten-2-one
CAS 625-33-2
CAS Min % 65.0
CAS Max % 100.0
Purity Grade Technical Grade
Assay ≥70.0%
Assay Percent Range 70%
Molecular Formula C5H8O
Linear Formula CH3CH=CHCOCH3
Formula Weight 84.12
MDL Number MFCD00009290
Accessories

GHS Signal Word: Danger

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic in contact with skin.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Highly flammable liquid and vapour.

SDS & Safety Documents