missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Penten-2-one, 70%, Tech., ACROS Organics™

Chemical Identifiers

CAS 625-33-2
CAS Min % 65.0
CAS Max % 100.0
Assay Percent Range 70%
Molecular Formula C5H8O
Linear Formula CH3CH=CHCOCH3
Formula Weight 84.12
MDL Number MFCD00009290
Chemical Name or Material 3-Penten-2-one
Assay ≥70.0%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC163290050 SDS Thermo Fisher Scientific
AC163290050
Glass bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC163290010 SDS Thermo Fisher Scientific
AC163290010
Glass bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC163290250 SDS Thermo Fisher Scientific
AC163290250
Glass bottle 25g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

625-33-2
100.0
C5H8O
84.12
3-Penten-2-one
Technical Grade
65.0
70%
CH3CH=CHCOCH3
MFCD00009290
≥70.0%

Specifications

Liquid
5g
0.86g/mL
100.0
Authentic
MFCD00009290
625-33-2
Glass bottle
≥70.0%
84.12
Clear colorless
02,306; 03,218; 04,371
65.0
CH3CH=CHCOCH3
C5H8O
70%
3-Penten-2-one
Technical Grade

Safety and Handling

GHS H Statement:
Toxic in contact with skin.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Highly flammable liquid and vapour.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger