Please login to your online account to display your discounted pricing

3-Penten-2-ol, 97%, Mixture of cis/trans (>90% trans), ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  150020250

 View more versions of this product

Catalog No. AC150020250

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Liquid
Formula Weight 86.13
Quantity 25g
Color Clear colorless
Beilstein 01, 443
TSCA TSCA
Assay 96% (trans+cis, approximately), 90% (trans-isomer, approximately)
Density 0.8430g/mL
EINECS Number 216-376-6
CAS Min % 96.0
CAS Max % 100.0
Chemical Name or Material 3-Penten-2-ol
Assay Percent Range 97%
Molecular Formula C5H10O
Linear Formula CH3CH=CHCH(OH)CH3
MDL Number MFCD00004544
CAS 1569-50-2
Health Hazard 1 Warning
Health Hazard 2 GHS P Statement Flammable liquid and vapor.
Health Hazard 3 GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Store in a well-ventilated place. Keep container tightly closed.
Infrared Spectrum Authentic
Packaging Glass bottle

Specifications

Chemical Name or Material 3-Penten-2-ol
CAS 1569-50-2
CAS Min % 96.0
CAS Max % 100.0
Assay 96% (trans+cis, approximately), 90% (trans-isomer, approximately)
Assay Percent Range 97%
Molecular Formula C5H10O
Linear Formula CH3CH=CHCH(OH)CH3
Formula Weight 86.13
MDL Number MFCD00004544
Accessories

Warning

GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Store in a well-ventilated place. Keep container tightly closed.

GHS P Statement Flammable liquid and vapor.

SDS & Safety Documents