missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Penten-2-ol, 97%, Mixture of cis/trans (>90% trans), ACROS Organics™

$233.28 - $619.59

Chemical Identifiers

CAS 1569-50-2
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C5H10O
Linear Formula CH3CH=CHCH(OH)CH3
Formula Weight 86.13
MDL Number MFCD00004544
Chemical Name or Material 3-Penten-2-ol
Assay 96% (trans+cis, approximately), 90% (trans-isomer, approximately)
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC150020250 SDS Thermo Fisher Scientific
150020250
Glass bottle 25g Each for $619.59
AC150020050 SDS Thermo Fisher Scientific
150020050
Glass bottle 5g Each for $233.28
Specifications Description & Specifications

Chemical Identifiers

1569-50-2
100.0
C5H10O
86.13
3-Penten-2-ol
96.0
97%
CH3CH=CHCH(OH)CH3
MFCD00004544
96% (trans+cis, approximately), 90% (trans-isomer, approximately)

Specifications

86.13
25g
01, 443
96.0
Authentic
Glass bottle
C5H10O
MFCD00004544
96% (trans+cis, approximately), 90% (trans-isomer, approximately)
Liquid
Clear colorless
0.8430g/mL
100.0
97%
CH3CH=CHCH(OH)CH3
1569-50-2
3-Penten-2-ol

Safety and Handling

GHS P Statement Flammable liquid and vapor.

GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Store in a well-ventilated place. Keep container tightly closed.

Warning

EINECSNumber : 216-376-6

TSCA : TSCA