missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Pentanone, 98%, pure, Acros Organics

Manufacturer:  Fisher Scientific Acros 155815000

 View more versions of this product

Catalog No. AC155815000



Specifications

Specifications

3-Pentanone
7°C
Glass bottle
0.815
100.0°C to 102.0°C
-40.0°C
98%
97.5
97.5% min. (GC)
C2H5COC2H5
01, IV, 3279
3-pentanone
FDPIMTJIUBPUKL-UHFFFAOYSA-N
pentan-3-one
7288
86.13
98%
0.8150g/mL
Authentic
1.3910 to 1.3930
0.47 mPa.s (20°C)
Colorless
500mL
96-22-0
100.0
C5H10O
MFCD00009320
15, 3137
Solubility in water: 50g/L (20°C). Other solubilities: soluble in alcohol and ether,slightly soluble in ethanol and acetone,miscible with many organic solvents
CCC(=O)CC
86.13
CHEBI:67886
Liquid
Pure

Chemical Identifiers

96-22-0
86.13
FDPIMTJIUBPUKL-UHFFFAOYSA-N
7288
pentan-3-one
C5H10O
MFCD00009320
3-pentanone
CHEBI:67886
CCC(=O)CC
Safety and Handling

Safety and Handling

GHS H Statement
Highly flammable liquid and vapour.
Causes serious eye irritation.
May cause respiratory irritation.
May cause drowsiness or dizziness.
Repeated exposure may cause skin dryness or cracking.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Avoid breathing d

GHS Signal Word: Danger

EINECSNumber : 202-490-3

RTECSNumber : SA8050000

TSCA : TSCA

SDS Documents