missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Pentanol, TCI America™

Manufacturer:  TCI America P005725ML

 View more versions of this product

Catalog No. P005725ML



Specifications

Specifications

3-Pentanol
Colorless
25mL
C5H12O
1105
AQIXEPGDORPWBJ-UHFFFAOYSA-N
pentan-3-ol
11428
88.15
≥98.0% (GC)
116°C
-8°C
584-02-1
MFCD00004574
3-pentanol, diethyl carbinol, sec-pentanol, pentanol-3, 3-pentyl alcohol, 3-hydroxypentane, unii-x4elc182i5, 1-ethyl-1-propanol, c2h5 2choh, isoamyl alcohol primary/secondary
CCC(CC)O
88.15
CHEBI:77519
Clear Liquid at 20°C

Chemical Identifiers

584-02-1
88.15
AQIXEPGDORPWBJ-UHFFFAOYSA-N
11428
pentan-3-ol
C5H12O
MFCD00004574
3-pentanol, diethyl carbinol, sec-pentanol, pentanol-3, 3-pentyl alcohol, 3-hydroxypentane, unii-x4elc182i5, 1-ethyl-1-propanol, c2h5 2choh, isoamyl alcohol primary/secondary
CHEBI:77519
CCC(CC)O
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0217

RTECSNumber : SA5075000

TSCA : Yes

Documents