Please login to your online account to display your discounted pricing

3-Pentanol, 98%, ACROS Organics™


Quantity 500g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC129995000

 View more versions of this product

Catalog No. AC129995000

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Formula Weight 88.15
Physical Form Liquid
Quantity 500g
Color Clear colorless
Density 0.815g/mL
Beilstein 01, 385
Merck Index 15, 7232
CAS Max % 100.0
Linear Formula C2H5CH(OH)C2H5
Infrared Spectrum Authentic
Refractive Index 1.4090 to 1.4110 (20°C, 589nm)
Chemical Name or Material 3-Pentanol
Assay Percent Range 98%
Molecular Formula C5H12O
CAS 584-02-1
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD00004574
CAS Min % 97.5
Packaging Glass bottle
Synonym Diethylecarbinol
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Harmful if inhaled.
May cause respiratory irritation.
Flammable liquid and vapor.

Specifications

Chemical Name or Material 3-Pentanol
CAS 584-02-1
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C5H12O
Linear Formula C2H5CH(OH)C2H5
Formula Weight 88.15
MDL Number MFCD00004574
Synonym Diethylecarbinol
Accessories

GHS Signal Word: Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Harmful if inhaled.
May cause respiratory irritation.
Flammable liquid and vapor.

SDS & Safety Documents