missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-(p-Tolyl)propionic Acid, TCI America™

Manufacturer:  TCI America T279725G

 View more versions of this product

Catalog No. T279725G



Specifications

Specifications

3-(p-Tolyl)propionic Acid
116°C
1505-50-6
MFCD00016561
LDYGRLNSOKABMM-UHFFFAOYSA-N
3-(4-methylphenyl)propanoic acid
73927
Crystal-Powder at 20°C
White-Yellow
25g
C10H12O2
3-4-methylphenyl propionic acid, 3-p-tolyl propionic acid, 3-p-tolyl propanoic acid, benzenepropanoic acid, 4-methyl, 3-4-methylphenyl propanoic acid, 4-methylphenylpropionic acid, 3-p-tolylpropanoic acid, p-methylhydrocinnamic acid, 4-methyl phenyl propionic acid, 4-methylhydrocinnamic acid
CC1=CC=C(C=C1)CCC(=O)O
164.204
164.20
≥98.0% (GC,T)

Chemical Identifiers

1505-50-6
164.204
LDYGRLNSOKABMM-UHFFFAOYSA-N
73927
CC1=CC=C(C=C1)CCC(=O)O
C10H12O2
MFCD00016561
3-4-methylphenyl propionic acid, 3-p-tolyl propionic acid, 3-p-tolyl propanoic acid, benzenepropanoic acid, 4-methyl, 3-4-methylphenyl propanoic acid, 4-methylphenylpropionic acid, 3-p-tolylpropanoic acid, p-methylhydrocinnamic acid, 4-methyl phenyl propionic acid, 4-methylhydrocinnamic acid
3-(4-methylphenyl)propanoic acid
Safety and Handling

Safety and Handling

TSCA : No

Documents