missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-(p-Tolyl)propionic acid, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC427360250

 View more versions of this product

Catalog No. AC427360250

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥96.0%
Crystalline Powder
White to yellow or pale orange
100.0
HOOC(CH2)2C6H4CH3
1505-50-6
MFCD00016561
3-(p-Tolyl)propionic acid
Authentic
164.2
25g
115° to 118°C
96.0
Consistent with structure
C10H12O2
97%
Glass bottle

Chemical Identifiers

1505-50-6
100.0
C10H12O2
164.2
3-(p-Tolyl)propionic acid
96.0
97%
HOOC(CH2)2C6H4CH3
MFCD00016561
≥96.0%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning