Please login to your online account to display your discounted pricing

3-(p-Tolyl)propionic acid, 97%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC427360050

 View more versions of this product

Catalog No. AC427360050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Crystalline Powder
Formula Weight 164.2
Assay ≥96.0%
Quantity 5g
Color White to yellow or pale orange
Melting Point 115° to 118°C
CAS Max % 100.0
CAS Min % 96.0
Linear Formula HOOC(CH2)2C6H4CH3
NMR Consistent with structure
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS 1505-50-6
Molecular Formula C10H12O2
MDL Number MFCD00016561
Assay Percent Range 97%
Chemical Name or Material 3-(p-Tolyl)propionic acid
Infrared Spectrum Authentic
Packaging Glass bottle

Specifications

Chemical Name or Material 3-(p-Tolyl)propionic acid
CAS 1505-50-6
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C10H12O2
Linear Formula HOOC(CH2)2C6H4CH3
Formula Weight 164.2
MDL Number MFCD00016561
Accessories

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents