Please login to your online account to display your discounted pricing

3-Oxopentanoic acid, 85%, Acros Organics


Quantity 1g

Manufacturer: thermo fisher scientific  AC452760010

 View more versions of this product

Catalog No. AC452760010

  • / Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Technical Grade
Formula Weight 116.12
Quantity 1g
Chemical Name or Material 3-Oxopentanoic acid
Assay Percent Range 85%
Molecular Formula C5H8O3
CAS 10191-25-0

Specifications

Chemical Name or Material 3-Oxopentanoic acid
CAS 10191-25-0
Purity Grade Technical Grade
Assay Percent Range 85%
Molecular Formula C5H8O3
Formula Weight 116.12