missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Oxopentanoic acid, 85%, Acros Organics

$166.82 - $166.82

Chemical Identifiers

CAS 10191-25-0
Assay Percent Range 85%
Molecular Formula C5H8O3
Formula Weight 116.12
Chemical Name or Material 3-Oxopentanoic acid
Grade Technical Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC452760010 Thermo Fisher Scientific
AC452760010
1g Each for $166.82
AC452760050 SDS Thermo Fisher Scientific
AC452760050
5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

10191-25-0
C5H8O3
3-Oxopentanoic acid
85%
116.12
Technical Grade

Specifications

10191-25-0
C5H8O3
1g
Technical Grade
85%
116.12
3-Oxopentanoic acid