Please login to your online account to display your discounted pricing

3-Oxopentanoic acid, 85%, Acros Organics


Quantity 5g

Manufacturer: thermo fisher scientific  AC452760050

 View more versions of this product

Catalog No. AC452760050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material 3-Oxopentanoic acid
CAS 10191-25-0
Purity Grade Technical Grade
Assay Percent Range 85%
Molecular Formula C5H8O3
Formula Weight 116.12
Quantity 5g

Specifications

Chemical Name or Material 3-Oxopentanoic acid
CAS 10191-25-0
Purity Grade Technical Grade
Assay Percent Range 85%
Molecular Formula C5H8O3
Formula Weight 116.12