Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Oxopentanenitrile, 99%

Chemical Identifiers

Chemical Name or Material 3-Oxopentanenitrile
CAS 33279-01-5
Molecular Formula C5H7NO
Formula Weight 97.12
MDL Number MFCD07367087
UN Number UN3439
Synonym Propionylacetonitrile
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5078903

  SDS
alfa aesar™
H5078903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH50789MD

  SDS
alfa aesar™
H50789MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 97.12
Chemical Name or Material 3-Oxopentanenitrile
Assay 99%
Molecular Formula C5H7NO
MDL Number MFCD07367087
CAS 33279-01-5
UN Number UN3439
Synonym Propionylacetonitrile
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H312-H332-H315-H319
Toxic if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 27° to 30°C
Quantity 1g
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

Applications
3-Oxopentanenitrile is used as pharmaceutical intermediates.

Notes
Incompatible with oxidizing agents. Store in a cool, dry, well-ventilated area.

Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H312-H332-H315-H319
Toxic if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.