Please login to your online account to display your discounted pricing

3-Oxo-3-[4-(trifluoromethoxy)phenyl]propanenitrile, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 3-Oxo-3-[4-(trifluoromethoxy)phenyl]propanenitrile
CAS 122454-46-0
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C10H6F3NO2
Linear Formula CF3OC6H4C(O)CH2CN
Formula Weight 229.16
MDL Number MFCD00052146
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

KM04838EA

 
fisher scientific company
KM04838EA
Amber Glass Bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 229.16
Quantity 10g
Assay 97%
CAS Min % 97.0
Chemical Name or Material 3-Oxo-3-[4-(trifluoromethoxy)phenyl]propanenitrile
Linear Formula CF3OC6H4C(O)CH2CN
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Causes serious eye irritation.
Causes skin irritation.
Harmful in contact with skin.
May cause respiratory irritation.
Assay Percent Range 97%
Molecular Formula C10H6F3NO2
MDL Number MFCD00052146
CAS 122454-46-0
Packaging Amber Glass Bottle
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.