Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Oxo-3,4-dihydro-2H-1,4-benzothiazine-2-acetic acid, 97%

$80.76 - $274.23

Chemical Identifiers

Chemical Name or Material 3-Oxo-3,4-dihydro-2H-1,4-benzothiazine-2-acetic acid
CAS 6270-74-2
Molecular Formula C10H9NO3S
Formula Weight 223.25
MDL Number MFCD00087524
Synonym 3,4-Dihydro-3-oxo-2H-1,4-benzothiazine-2-acetic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2782406

  SDS
alfa aesar™
H2782406
5g Each for $80.76

AAH2782414

  SDS
alfa aesar™
H2782414
25g Each for $274.23
Description & Specifications

Specifications

Formula Weight 223.25
Chemical Name or Material 3-Oxo-3,4-dihydro-2H-1,4-benzothiazine-2-acetic acid
CAS 6270-74-2
Synonym 3,4-Dihydro-3-oxo-2H-1,4-benzothiazine-2-acetic acid
Health Hazard 1 Warning
Molecular Formula C10H9NO3S
Quantity 5g
MDL Number MFCD00087524
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 97%
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 185° to 190°C
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.