Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Oxo-2,3-dihydro-4H-1,4-benzoxazine-4-acetic acid, 98%

$195.49 - $694.22

Chemical Identifiers

Chemical Name or Material 3-Oxo-2,3-dihydro-4H-1,4-benzoxazine-4-acetic acid
CAS 26494-55-3
Molecular Formula C10H9NO4
Formula Weight 207.19
MDL Number MFCD00269587
Synonym 2,3-Dihydro-3-oxo-4H-1,4-benzoxazine-4-acetic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5027503

  SDS
alfa aesar™
H5027503
1g Each for $694.22

AAH50275MD

  SDS
alfa aesar™
H50275MD
250mg Each for $195.49
Description & Specifications

Specifications

Formula Weight 207.19
Chemical Name or Material 3-Oxo-2,3-dihydro-4H-1,4-benzoxazine-4-acetic acid
Assay 98%
Molecular Formula C10H9NO4
MDL Number MFCD00269587
CAS 26494-55-3
Synonym 2,3-Dihydro-3-oxo-4H-1,4-benzoxazine-4-acetic acid
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.