missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-Oxetanone, 95%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC451200050
Specifications
3-Oxetanone | |
140°C | |
6704-31-0 | |
C3H4O2 | |
ROADCYAOHVSOLQ-UHFFFAOYSA-N | |
oxetan-3-one | |
15024254 | |
95% |
26°C | |
5g | |
94% min. (GC) | |
1,3-epoxy-2-propanone, 1,3-epoxypropanone, 3-oxentanone, 3-oxetanone, 3-oxooxetane, acmc-209nxj, ksc358e8n, oxetan-3-on, oxetane-3-one, pubchem15880 | |
C1C(=O)CO1 | |
72.06 | |
72.06 |
Chemical Identifiers
6704-31-0 | |
72.06 | |
1,3-epoxy-2-propanone, 1,3-epoxypropanone, 3-oxentanone, 3-oxetanone, 3-oxooxetane, acmc-209nxj, ksc358e8n, oxetan-3-on, oxetane-3-one, pubchem15880 | |
oxetan-3-one |
C3H4O2 | |
ROADCYAOHVSOLQ-UHFFFAOYSA-N | |
15024254 | |
C1C(=O)CO1 |
Safety and Handling
GHS H Statement
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS Signal Word: Warning