Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Oxetanemethanol, 96%

$1,145.14 - $1,145.14

Chemical Identifiers

Chemical Name or Material 3-Oxetanemethanol
CAS 6246-06-6
Molecular Formula C4H8O2
Formula Weight 88.11
MDL Number MFCD17015894
Synonym 3-(Hydroxymethyl)oxetane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6633303

  SDS
alfa aesar™
H6633303
1g Each for $1,145.14

AAH66333MD

  SDS
alfa aesar™
H66333MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 88.11
Physical Form Liquid
Assay 96%
MDL Number MFCD17015894
CAS 6246-06-6
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Color Colorless to pale yellow
Chemical Name or Material 3-Oxetanemethanol
Molecular Formula C4H8O2
Synonym 3-(Hydroxymethyl)oxetane
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H318-H302-H335-H315
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Storage Note 1 -30° to -10°C
Flash Point 106°C (223°F)
Quantity 1g
Sensitivity Moisture Sensitive
Recommended Storage −30° to −10°C
Safety and Handling

Danger

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H318-H302-H335-H315
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.

−30° to −10°C

-30° to -10°C