missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Oxabicyclo[3.1.0]hexane-2,4-dione, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 370120010

 View more versions of this product

Catalog No. AC370120010



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3-Oxabicyclo[3.1.0]hexane-2, 4-dione
5617-74-3
100.0
White
Authentic
Glass bottle
MFCD00126929
3-oxabicyclo 3.1.0 hexane-2,4-dione, 1,2-cyclopropanedicarboxylic anhydride, oxabicyclohexanedione, pubchem18320, cyclopropane-1,2-dicarboxylic anhydride, 2,4-dioxo-3-oxabicyclo 3.1.0 hexane, 3-oxa-bicyclo 3.1.0 hexane-2,4-dione, 3-oxabicyclo 3.1.0-hexane-2,4-dione, 3-oxa-bicyclo 3.1.0-hexane-2,4-dione
ZRMYHUFDVLRYPN-UHFFFAOYSA-N
3-oxabicyclo[3.1.0]hexane-2,4-dione
2734721
98%
98%
97.5
54°C to 60°C
100°C to 102°C
97.5% min. (GC)
C5H4O3
1 g
Solubility in water: insoluble
C1C2C1C(=O)OC2=O
112.09
112.09
Powder, Crystals or Chunks

Chemical Identifiers

5617-74-3
112.09
ZRMYHUFDVLRYPN-UHFFFAOYSA-N
2734721
C1C2C1C(=O)OC2=O
C5H4O3
MFCD00126929
3-oxabicyclo 3.1.0 hexane-2,4-dione, 1,2-cyclopropanedicarboxylic anhydride, oxabicyclohexanedione, pubchem18320, cyclopropane-1,2-dicarboxylic anhydride, 2,4-dioxo-3-oxabicyclo 3.1.0 hexane, 3-oxa-bicyclo 3.1.0 hexane-2,4-dione, 3-oxabicyclo 3.1.0-hexane-2,4-dione, 3-oxa-bicyclo 3.1.0-hexane-2,4-dione
3-oxabicyclo[3.1.0]hexane-2,4-dione
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only